Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPD52L3/D55 Antibody (OTI8C12), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274567V
Description
TPD52L3/D55 Monoclonal specifically detects TPD52L3/D55 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
TPD52L3/D55 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TPD52L3 | |
Full length recombinant protein of human TPD52L3/D55 (NP_001001875) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI8C12 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
hD55, MGC26757, NYDSP25, NYD-SP25, protein kinase NYD-SP25, Testis development protein NYD-SP25, tumor protein D52-like 3MGC45374, tumor protein D55 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
89882.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction