Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPD52L3/D55 Antibody (OTI8C12), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274567MFV610
Description
TPD52L3/D55 Monoclonal antibody specifically detects TPD52L3/D55 in Human samples. It is validated for Flow CytometrySpecifications
TPD52L3/D55 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI8C12 | |
Flow Cytometry | |
hD55, MGC26757, NYDSP25, NYD-SP25, protein kinase NYD-SP25, Testis development protein NYD-SP25, tumor protein D52-like 3MGC45374, tumor protein D55 | |
Full length human recombinant protein of human TPD52L3(NP_001001875) produced in HEK293T cell. | |
0.1 mL | |
Protein Kinase | |
89882 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction