Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPD52L3 Monoclonal Antibody (OTI3D11)
GREENER_CHOICE

Catalog No. PIMA526023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526023 Supplier Invitrogen™ Supplier No. MA526023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPD52L3 Monoclonal Antibody for Western Blot, IHC (P)

Belongs to the TPD52 family and is specifically expressed in testis. Expressed at 5.6-fold higher levels in adult testis than in fetal testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPD52L3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TPD52L3
Gene Accession No. Q96J77
Gene Alias d55; hD55; NYDSP25; protein kinase NYD-SP25; RGD1309391; Testis development protein NYD-SP25; testis tissue sperm-binding protein Li 87P; Tpd52l3; Trpd52l3; Tumor protein D52-like 3; tumor protein D55
Gene Symbols TPD52L3
Host Species Mouse
Immunogen Full length human recombinant protein of TPD52L3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89882
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.