Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPD52L3 Monoclonal Antibody (OTI4C12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504965S


Description
TPD52L3 Monoclonal antibody specifically detects TPD52L3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TPD52L3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P16284 | |
Mouse | |
Affinity chromatography | |
RUO | |
5175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin) | |
OTI4C12 | |
Unconjugated | |
Pecam1 | |
d55; hD55; NYDSP25; protein kinase NYD-SP25; RGD1309391; Testis development protein NYD-SP25; testis tissue sperm-binding protein Li 87P; Tpd52l3; Trpd52l3; Tumor protein D52-like 3; tumor protein D55 | |
Full length human recombinant protein of human PECAM1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction