Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPD52L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535291
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535291 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535291 Supplier Invitrogen™ Supplier No. PA535291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPD52L3 Polyclonal Antibody for Western Blot, ICC/IF

Belongs to the TPD52 family and is specifically expressed in testis. Expressed at 5.6-fold higher levels in adult testis than in fetal testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPD52L3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TPD52L3
Gene Accession No. Q96J77
Gene Alias d55; hD55; NYDSP25; protein kinase NYD-SP25; RGD1309391; Testis development protein NYD-SP25; testis tissue sperm-binding protein Li 87P; Tpd52l3; Trpd52l3; Tumor protein D52-like 3; tumor protein D55
Gene Symbols TPD52L3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 10-38 amino acids from the N-terminal region of human TPD52L3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.