Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPGS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566420 Supplier Invitrogen™ Supplier No. PA566420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQLTQSSMYSL PNAPTLADLE DDTHEASDDQ PEKPHFDSRS VIFELDSCNG SGKVCLVYKS GKPALAEDTE IWFLDRALYW HFL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 81%.

This gene encodes a protein that is a component of the neuronal polyglutamylase complex, which plays a role in post-translational addition of glutamate residues to C-terminal tubulin tails. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPGS2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TPGS2
Gene Accession No. Q68CL5
Gene Alias C18orf10; HMFN0601; HsT3006; L17; PGs2; TPGS2; Tubulin polyglutamylase complex subunit 2
Gene Symbols TPGS2
Host Species Rabbit
Immunogen Recombinant Human TPGS2. Recombinant protein control fragment (Product #RP-105206).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.