Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589881
Description
Positive Samples: U-87MG, SH-SY5Y, Mouse brain, Rat brain Immunogen sequence: FDPKTTCLQE CLITTFQEAY FVSESFEEAK EKMRDFAKSI TRPFSVYFNP YTQSIEILKD TRSIENVVQD.
Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
Specifications
| TPH2 | |
| Polyclonal | |
| Unconjugated | |
| Tph2 | |
| ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 121278, 216343, 317675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q8CGU9, Q8CGV2, Q8IWU9 | |
| Tph2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-70 of human TPH2 (NP_7754892). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction