Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142777
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142777 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142777 Supplier Invitrogen™ Supplier No. PA5142777
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPH2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Tph2
Gene Accession No. Q8IWU9
Gene Alias ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Gene Symbols Tph2
Host Species Goat
Immunogen Peptide with sequence RGFSLDSAVPEEHQ-C.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121278
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.