Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPI1 Monoclonal Antibody (P1111)

Catalog No. PIMA518284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518284 Supplier Invitrogen™ Supplier No. MA518284
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPI1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone P1111
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Tpi1
Gene Accession No. P17751, P48500, P60174
Gene Alias AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C
Gene Symbols Tpi1
Host Species Mouse
Immunogen Recombinant fragment corresponding to a region within amino acids 187 and 249 of Triosephosphate isomerase.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21991, 24849, 7167
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.