Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598317
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598317 Supplier Invitrogen™ Supplier No. PA598317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPI1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Tpi1
Gene Accession No. P17751, P60174
Gene Alias AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C
Gene Symbols Tpi1
Host Species Rabbit
Immunogen Recombinant Human Triosephosphate isomerase protein (169-228AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21991, 7167
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.