Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPI1 Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142133
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise reported isoforms 1 and 2 (NP_000356.1; NP_001152759.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
Specifications
TPI1 | |
Polyclonal | |
Biotin | |
Tpi1 | |
AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
21991, 7167 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P17751, P60174 | |
Tpi1 | |
Peptide with sequence C-LKPEFVDIINAKQ (aa 237-249). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction