Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527970
Description
Recommended positive controls: H1299, mouse liver. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPK1 is a protein, that exists as a homodimer, which catalyzes the conversion of thiamine to thiamine pyrophosphate.
Specifications
TPK1 | |
Polyclonal | |
Unconjugated | |
Tpk1 | |
HTPK1; mTPK1; placental protein 20; PP20; Thiamin pyrophosphokinase 1; thiamine diphosphokinase; thiamine kinase; thiamine pyrophosphokinase; thiamine pyrophosphokinase 1; THMD5; TPK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27010, 29807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.96 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9H3S4, Q9R0M5 | |
Tpk1 | |
Recombinant fragment corresponding to a region within amino acids 4 and 243 of Human TPK1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction