Learn More
Invitrogen™ TPK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596297
Description
Immunogen sequence: MEHAFTPLEP LLSTGNLKYC LVILNQPLDN YFRHLWNKAL LRACADGGAN RLYDITEGER ESFLPEFING DFDSIRPEVR EYYATKGCEL ISTPDQDHTD FTKCLKMLQK KIEEKDLKVD VIVTLGGLAG RFDQIMASVN TLFQATHITP FPIIIIQEES LIYLLQPGKH RLHVDTGMEG DWCGLIPVGQ PCMQVTTTGL KWNLTNDVLA FGTLVSTSNT YDGSGVVTVE TDHPLLWTMA IKS; Positive Samples: HepG2, SKOV3, Mouse kidney, Mouse liver, Rat liver.
TPK1 is a protein, that exists as a homodimer, which catalyzes the conversion of thiamine to thiamine pyrophosphate.
Specifications
TPK1 | |
Polyclonal | |
Unconjugated | |
Tpk1 | |
HTPK1; mTPK1; placental protein 20; PP20; Thiamin pyrophosphokinase 1; thiamine diphosphokinase; thiamine kinase; thiamine pyrophosphokinase; thiamine pyrophosphokinase 1; THMD5; TPK1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27010, 29807, 680668 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H3S4, Q9R0M5 | |
Tpk1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-243 of human TPK1 (NP_0718902). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.