Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533914
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533914 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533914 Supplier Invitrogen™ Supplier No. PA533914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Elephant, Bovine, Dog, Bat (100%) Mouse, Rat, Hamster, Panda, Horse (94%).

MAP3K8 (COT) is a serine/threonine kinase which can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle. Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK8 is able to activate NF-kappa-B 1 by stimulating proteasome-mediated proteolysis of NF-kappa-B 1/p105. This cytoplasmic protein is expressed in several normal tissues and human tumor-derived cell lines. The 58 kDa form is activated specifically during the S and G2/M phases of the cell cycle. The longer form undergoes phosphorylation on Ser residues mainly, and the shorter form on both Ser and Thr residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MAP3K8
Gene Accession No. P41279
Gene Alias augmented in rheumatoid arthritis 2; AURA2; Cancer Osaka thyroid oncogene; cancer Osaka thyroid, oncogene; CCOT; c-COT; COT; cot (cancer Osaka thyroid) oncogene; COT proto-oncogene serine/threonine-protein kinase; Cot/Tpl2; D17TPL2; EC 1.14.16.2; Est; Estf; Ewing sarcoma transformant; FLJ10486; M3K8; MAP3K 8; Map3k8; MEKK8; mitogen activated protein kinase kinase kinase 8; mitogen-activated protein kinase kinase kinase 8; Protein kinase cot; Proto-oncogene c-Cot; proto-oncogene serine/threoine protein kinase; serine/threonine kinase (Tpl-2); serine/threonine-protein kinase cot; TH isoform 3; TH isoform a; TH-4; TPL 2; Tpl2; TPL-2; Tumor progression locus 2; TY3H; TYH
Gene Symbols MAP3K8
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from near C-terminus of human MAP3K8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1326, 487085, 535622
Target Species Human, Canine, Monkey, Bovine, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.