Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584164
Description
Immunogen sequence: LNRRIQLVEE ELDRAQERLA TALQKLEE.
Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.
Specifications
TPM1 | |
Polyclonal | |
Unconjugated | |
TPM1 | |
AA986836; AI854628; alpha tropomyosin; alpha tropomyosin of brain; alpha tropomyosin of fibroblasts F2; alpha-fTM; Alpha-tm; alpha-Tm-1; alpha-Tm-2 isoform; alpha-tropomyosin; alpha-tropomyosin (partial); alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; C15orf13; cardiac alpha tropomyosin; cardiomyopathy, hypertrophic 3; CMD1Y; CMH3; epididymis secretory protein Li 265; HEL-S-265; hepatoma alpha tropomyosin; HTM-alpha; LVNC9; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; Tm1; TM2; Tm3; Tma2; Tmpa; Tmsa; Tpm1; Tpm-1; tpm1 protein; TPM1-1; TPM1-alpha; TPMA; tropomyosin; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1; tropomyosin 1 (alpha); tropomyosin 1 alpha chain; tropomyosin 1, alpha; tropomyosin 1-1; tropomyosin 3 alpha; tropomyosin alpha-1 chain; Tropomyosin-1; unnamed protein product; zgc:171719; zgc:55951; zgc:77009 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7168 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09493 | |
TPM1 | |
Recombinant protein corresponding to Human TPM1. Recombinant protein control fragment (Product #RP-104195). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction