Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543143
Description
Peptide sequence: AETRAEFAER SVAKLEKTID DLEDEVYAQK MKYKAISEEL DNALNDITSL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 85%; Human: 100%; Mouse: 100%; Pig: 85%; Rabbit: 100%; Rat: 100%; Sheep: 85%; Zebrafish: 86%.
TPM2 (tropomyosin 2) is beta-tropomyosin, an isoform of tropomyosin that is mainly expressed in slow, type 1 muscle fibers. TPM2 binds to actin filaments in muscle and non-muscle cells. Plays a central role in association with the troponin complex in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. Mutations in this gene can cause nemaline myopathy 4, arthrogryposis dital 1A/2B, and cap myopathy 2.
Specifications
| TPM2 | |
| Polyclonal | |
| Unconjugated | |
| TPM2 | |
| AMCD1; beta tropomyosin; beta-tropomyosin; cb836; cytoskeletal protein; DA1; DA2B; epididymis secretory protein Li 273; fj63f03; HEL-S-273; NEM4; nemaline myopathy type 4; RP11-112J3.4; TMSB; TPM2; Tpm-2; tpm4l; Trop-2; tropomyosin; tropomyosin 2; tropomyosin 2 (beta); tropomyosin 2, beta; tropomyosin beta chain; Tropomyosin-2; unnamed protein product; wu:fj63f03; zgc:86810 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7169 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P07951 | |
| TPM2 | |
| Synthetic peptide directed towards the C-terminal of human TPM2 (aa 235-284). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction