Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591925 Supplier Invitrogen™ Supplier No. PA591925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SW620, HeLa, MCF7, U-87MG, Mouse skeletal muscle, Mouse lung; Cellular Location: Cytoplasm, cytoskeleton Immunogen sequence: MDAIKKKMQM LKLDKENAID RAEQAEADKK QAEDRCKQLE EEQQALQKKL KGTEDEVEKY SESVKEAQEK LEQAEKKATD AEADVASLNR RIQLVEEELD.

TPM2 (tropomyosin 2) is beta-tropomyosin, an isoform of tropomyosin that is mainly expressed in slow, type 1 muscle fibers. TPM2 binds to actin filaments in muscle and non-muscle cells. Plays a central role in association with the troponin complex in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. Mutations in this gene can cause nemaline myopathy 4, arthrogryposis dital 1A/2B, and cap myopathy 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPM2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TPM2
Gene Accession No. P07951, P58774, P58775
Gene Alias AMCD1; beta tropomyosin; beta-tropomyosin; cb836; cytoskeletal protein; DA1; DA2B; epididymis secretory protein Li 273; fj63f03; HEL-S-273; NEM4; nemaline myopathy type 4; RP11-112J3.4; TMSB; TPM2; Tpm-2; tpm4l; Trop-2; tropomyosin; tropomyosin 2; tropomyosin 2 (beta); tropomyosin 2, beta; tropomyosin beta chain; Tropomyosin-2; unnamed protein product; wu:fj63f03; zgc:86810
Gene Symbols TPM2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22004, 500450, 7169
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.