Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPM2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982244
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2244 Supplier Bioss Supplier No. BS1243R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPM2 (tropomyosin 2) is beta-tropomyosin, an isoform of tropomyosin that is mainly expressed in slow, type 1 muscle fibers. TPM2 binds to actin filaments in muscle and non-muscle cells. Plays a central role in association with the troponin complex in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. Mutations in this gene can cause nemaline myopathy 4, arthrogryposis dital 1A/2B, and cap myopathy 2.

Specifications

Antigen TPM2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TPM2
Gene Accession No. P07951
Gene Alias AMCD1; beta tropomyosin; Beta-tropomyosin; BOS_8893; cb836; DA1; DA2B; epididymis secretory protein Li 273; fj63f03; HEL-S-273; LOC100125984; NEM4; nemaline myopathy type 4; RP11-112J3.4; TMSB; Tpm2; Tpm-2; tpm4l; Trop-2; tropomyosin 2; tropomyosin 2 (beta); tropomyosin 2, beta; Tropomyosin beta chain; Tropomyosin-2; wu:fj63f03; zgc:86810
Gene Symbols TPM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TPM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7169
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.