Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529005 Supplier Invitrogen™ Supplier No. PA529005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (83%), Dog (93%), Pig (100%), Chicken (84%), Rhesus Monkey (93%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TPM3 (tropomyosin 3) is a member of the tropomyosin family of actin-binding proteins involved in the contractile system of striated and smooth muscles and the cytoskeleton of non-muscle cells. Tropomyosins are dimers of coiled-coil proteins that polymerize end-to-end along the major groove in most actin filaments. They provide stability to the filaments and regulate access of other actin-binding proteins. In muscle cells, they regulate muscle contraction by controlling the binding of myosin heads to the actin filament. Mutations in this gene result in autosomal dominant nemaline myopathy, and oncogenes formed by chromosomal translocations involving this locus are associated with cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPM3
Applications Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TPM3
Gene Accession No. P06753
Gene Alias alpha-tropomyosin, slow skeletal; CAPM1; CFTD; cytoskeletal tropomyosin TM30; epididymis luminal protein 189; epididymis secretory sperm binding protein Li 82p; gamma-TM; gamma-tropomyosin; heat-stable cytoskeletal protein 30 kDa; HEL-189; HEL-S-82p; hscp30; hTM30nm; hTM5; hTMnm; NEM1; nonmuscle tropomyosin 5; OK/SW-cl.5; skalphaTM.2; TM3; TM30; TM30nm; TM5; TM-5; Tm5NM; TMnm; TPM3; Tpm5; Tpm-5; TPMsk3; TRK; Trop-5; tropomyosin; tropomyosin 3; tropomyosin 3, gamma; tropomyosin 5; tropomyosin alpha-3 chain; tropomyosin gamma; tropomyosin non-muscle; tropomyosin-3; Tropomyosin-5
Gene Symbols TPM3
Host Species Rabbit
Immunogen Full length human Tropomyosin 3 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.