Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPMT Antibody (2E7), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197939C
Description
TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
TPMT | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TPMT | |
Recombinant human TPMT (1-245aa) purified from E. coli | |
100 μL | |
Proteases & Other Enzymes, Signal Transduction | |
7172 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
20000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human TPMT mAb, clone 2E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TPMT protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction