Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPMT Antibody (SR1518), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TPMT |
---|---|
Clone | SR1518 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Description
TPMT Monoclonal antibody specifically detects TPMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
TPMT | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
A synthesized peptide derived from human TPMT (Uniprot #: P51580) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1518 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Proteases & Other Enzymes, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7172 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title