Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587693 Supplier Invitrogen™ Supplier No. PA587693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDGTRTSLDI EEYSDTEVQK NQVLTLEEWQ DKWVNGKTAF HQEQGHQLLK KHLDTFLKGK SGLRVFFPLC GKAVEMKWFA DRGHSVVGVE ISELGIQEFF TEQNLSYSEE PITEIPGTKV FKSSSGNISL YCCSIFDLPR TNIGKFDMIW DRGALVAINP GDRKCYADTM FSLLGKKFQY LLCVLSYDPT KHPGPPFYVP HAEIERLFGK ICNIRCLEKV DAFEERHKSW GIDCLFEKLY LLTEK; Positive Samples: K562; Cellular Location: Cytoplasm.

Catalyzes the S-methylation of thiopurine drugs such as 6-mercaptopurine. Defects in TPMT are the cause of thiopurine S-methyltransferase deficiency (TPMT deficiency). TPMT is an enzyme involved in the normal metabolic inactivation of thiopurine drugs. These drugs are generally used as immunosupressants or cytotoxic drugs and are prescribed for a variety of clinical conditions including leukemia, autoimmune disease and organ transplantation. Patients with intermediate or no TPMT activity are at risk of toxicity after receiving standard doses of thiopurine drugs and it is shown that inter-individual differences in response to these drugs are largely determined by genetic variation at the TPMT locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPMT
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TPMT
Gene Accession No. P51580
Gene Alias AW106912; HGNC:12014; S-adenosyl-L-methionine:thiopurine S-methyltransferase; thiopurine methyltransferase; Thiopurine S-methyltransferase; TPMT; TPMTD; truncated thiopurine methyltransferase
Gene Symbols TPMT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-245 of human TPMT (NP_0003581).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7172
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.