Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPO Rabbit anti-Human, Unconjugated, Clone: AFFI-20, Abnova™

Catalog No. 89339754 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-754 Supplier Abnova Corporation Supplier No. MAB22016
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TPO Monoclonal antibody specifically detects TPO in Human samples. It is validated for WB, IP.

Specifications

Antigen TPO
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone AFFI-20
Conjugate Unconjugated
Description thyroid peroxidase
Gene TPO
Gene Alias MSA, TPX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human TPO
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.