Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPP1 Mouse anti-Human, Clone: 2E12, MilliporeSigma™

Catalog No. MABN1806MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1806MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1806MI Supplier MilliporeSigma Supplier No. MABN1806
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TPP1, clone 2E12 is a highly specific mouse monoclonal antibody, that targets TPP1/Tripeptidyl-peptidase 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2E12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TPP1,CLN2,GIG1,UNQ267/PRO304
Host Species Mouse
Immunogen A recombinant fragment corresponding to amino acids 195-563 of human TPP1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000382.3
Test Specificity Clone 2E12 specifically immunostains tripeptidyl peptidase 1 in human placenta, heart, and pancreas.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.