Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110512 Supplier Invitrogen™ Supplier No. PA5110512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATAATEEPF PFHGLLPKKE TGAASFLCRY PEYDGRGVLI AVLDTGVDPG APGMQVTTDG KPKIVDIIDT TGSGDVNTAT EVEPKDGEIV GLSGRVLKIP ASWTNPSGKY HIGIKNGYDF YPKALKERIQ KERKEKIWDP VHRVALAEAC RKQEEFDVAN NGSSQANKLI KEELQSQVEL LNSFEKKYSD PGPVYDCLVW HDGEVWRACI DSNEDGDLSK STVLRNYKEA QEYGSFGTAE MLNYSVNIYD DGNLLSIVTS GGAHGTHVAS IAAGHFPEEP ERNGVAPGAQ ILSIKIGDTR.

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TPP2
Gene Accession No. P29144, Q64514, Q64560
Gene Alias TPP2; TPP-2; TppII; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase 2; tripeptidyl peptidase II; tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
Gene Symbols TPP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TPP2 (NP_003282.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22019, 7174, 81815
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.