Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPPP/p25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18096225UL
Description
TPPP/p25 Polyclonal specifically detects TPPP/p25 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TPPP/p25 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
O94811 | |
TPPP | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SLESEGAGEGAAASPELSALEEAFRRFAVHGDARATGREMHGKNWSKLC | |
Affinity Purified | |
RUO | |
11076 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
p24, P24,25 kDa brain-specific protein, P25, p25alpha, p25-alpha, TPPP/p25brain specific protein p25 alpha, TPPP1glycogen synthase kinase 3 (GSK3) inhibitor p24, tubulin polymerization promoting protein, tubulin polymerization-promoting protein | |
Rabbit | |
26 kDa | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction