Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPPP/p25 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP149833
Description
TPPP/p25 Polyclonal specifically detects TPPP/p25 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
TPPP/p25 | |
Polyclonal | |
Western Blot 0.0001-0.01 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:32000 | |
NP_008961.1 | |
TPPP | |
Peptide with sequence C-DPSKDRAAKRLSLES corresponding to internal region according to NP_008961.1. | |
0.1 mg | |
Primary | |
This antibody is specific to the Internal region. Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Pig. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
p24, P24,25 kDa brain-specific protein, P25, p25alpha, p25-alpha, TPPP/p25brain specific protein p25 alpha, TPPP1glycogen synthase kinase 3 (GSK3) inhibitor p24, tubulin polymerization promoting protein, tubulin polymerization-promoting protein | |
Goat | |
Affinity Purified | |
RUO | |
11076 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction