Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPPP2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297003V
Description
TPPP2 Polyclonal specifically detects TPPP2 in Human samples. It is validated for ELISA.Specifications
TPPP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
122664 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 405 | |
C14orf8, chromosome 14 open reading frame 8, P18, p25beta, Protein p25-beta, TPPP/p18, tubulin polymerization-promoting protein family member 2 | |
Produced in rabbits immunized with purified, recombinant TPPP2 (AAH38970.1; Met1-Lys170). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction