Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPPP2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297873PE
Description
TPPP2 Polyclonal antibody specifically detects TPPP2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
TPPP2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
C14orf8, chromosome 14 open reading frame 8, P18, p25beta, Protein p25-beta, TPPP/p18, tubulin polymerization-promoting protein family member 2 | |
Produced in rabbits immunized with E. coli-derived Human TPPP2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
122664 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction