Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPPP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112776
Description
TPPP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
| TPPP2 | |
| Polyclonal | |
| Unconjugated | |
| TPPP2 | |
| C14orf8; CT152; Gm1790; Gm77; P18; p25 beta; p25beta; Protein p25-beta; RGD1566185; TPPP/p18; TPPP2; tubulin polymerization promoting protein family member 2; tubulin polymerization-promoting protein family member 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 122664 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P59282 | |
| TPPP2 | |
| Recombinant Human Tubulin polymerization-promoting protein family member 2 protein (1-170AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction