Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPPP3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300282F
Description
TPPP3 Polyclonal specifically detects TPPP3 in Human samples. It is validated for ELISA.Specifications
TPPP3 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
51673 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
brain specific protein, CGI-38, p20, p25gamma, TPPP/p20, tubulin polymerization-promoting protein family member 3 | |
Produced in rabbits immunized with purified, recombinant TPPP3 (Q9BW30; Met1-Lys176). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction