Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPPP3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561560 Supplier Thermo Scientific Supplier No. PA561560

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: ATKRFKGKSK EEAFDAICQL VAGKEPANVG VTKAKTGGAV DRL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPPP3
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tppp3
Gene Accession No. Q9BW30
Gene Alias 2700055K07Rik; brain specific protein; Ceacam9; CGI-38; hypothetical protein LOC614988; mmCGM8; p20; p25gamma; RGD1305061; TPPP/p20; TPPP3; tubulin polymerization promoting protein family member 3; tubulin polymerization-promoting protein family member 3
Gene Symbols Tppp3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPPP3
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.