Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555054 Supplier Invitrogen™ Supplier No. PA555054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRPPHPLPPR LTIHAPPQEL GPPVQRIQMT RRQSVGRGLQ LTPGIGGMQQ HFFDDEDRTV PSTPTLVVPH RTDGFAEAIH SPQVAGVPRF RFGPPEDMPQ TSSSHS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPR
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TPR
Gene Accession No. P12270
Gene Alias 2610029M07Rik; C77892; Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein Tpr; nucleoprotein TPR; nucleoprotein TPR-like protein; Tpr; translocated promoter region (to activated MET oncogene); translocated promoter region and nuclear basket protein; Translocated promoter region protein; translocated promoter region, nuclear basket protein; tumor potentiating region
Gene Symbols TPR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPR. Recombinant protein control fragment (Product #RP-91829).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.