Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPRA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534274
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Rabbit (100%) Dog, Bovine, Elephant, Panda, Horse (94%).
TPRA1 gene ontology (GO) annotations include aging; embryonic cleavage; lipid metabolic process; negative regulation of mitotic cell cycle phase transition.
Specifications
| TPRA1 | |
| Polyclonal | |
| Unconjugated | |
| TPRA1 | |
| 40kDa; FLJ32197; G protein-coupled receptor 175; GPR175; Integral membrane protein GPR175; PP6566; seven transmembrane domain orphan receptor; TMEM227; TPRA1; Tpra40; Transmembrane domain protein of 40 kDa regulated in adipocytes; transmembrane domain protein regulated in adipocytes 40 kDa; transmembrane protein 227; transmembrane protein adipocyte associated 1; transmembrane protein adipocyte-associated 1; transmembrane protein, adipocyte asscociated 1; transmembrane protein, adipocyte associated 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100359294, 131601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q86W33 | |
| TPRA1 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human TPRA1 / GPR175. | |
| 50 μg | |
| Primary | |
| Human, Rabbit, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction