Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPRA1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132706
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132706 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132706 Supplier Thermo Scientific Supplier No. PA132706

Rabbit Polyclonal Antibody

PA1-32706 detects GPCR LOC51210 from human samples.

TPRA1 gene ontology (GO) annotations include aging; embryonic cleavage; lipid metabolic process; negative regulation of mitotic cell cycle phase transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPRA1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TPRA1
Gene Accession No. Q86W33
Gene Alias 40kDa; FLJ32197; G protein-coupled receptor 175; GPR175; Integral membrane protein GPR175; PP6566; seven transmembrane domain orphan receptor; TMEM227; TPRA1; Tpra40; Transmembrane domain protein of 40 kDa regulated in adipocytes; transmembrane domain protein regulated in adipocytes 40 kDa; transmembrane protein 227; transmembrane protein adipocyte associated 1; transmembrane protein adipocyte-associated 1; transmembrane protein, adipocyte asscociated 1; transmembrane protein, adipocyte associated 1
Gene Symbols TPRA1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human GPR175
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.