Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPRBK Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568258
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8258 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8258 Supplier Bethyl Laboratories, Inc Supplier No. A304636AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TPRBK may be involved in the condensation of spindle midzone microtubules, leading to the formation of midbody. Diseases associated with this gene include Cleft Soft Palate.

Specifications

Antigen TPRBK
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TTC28
Gene Accession No. Q80XJ3, Q96AY4
Gene Alias KIAA1043, TPR repeat protein 28, TPR repeat-containing big gene cloned at Keio, TPRBK, TPR-containing big gene cloned at Keio
Gene Symbols TTC28
Host Species Rabbit
Immunogen Between 2450 and 2500
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209683, 23331
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.