Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPRG1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534885 Supplier Invitrogen™ Supplier No. PA534885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, Neuro2A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Presynaptic protein involved in the synaptic transmission tuning. Regulates synaptic release probability by decreasing the calcium sensitivity of release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPRG1L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene TPRG1L
Gene Accession No. Q5T0D9, Q9DBS2
Gene Alias 1200015A19Rik; Fam79a; family with sequence similarity 79, member A; h-mover; mossy fiber terminal-associated vertebrate-specific presynaptic protein; mossy-fiber terminal-associated vertebrate-specific presynaptic protein; Mover; Protein FAM79A; RP11-46F15.3; Svap30; synaptic vesicle associated protein of 30 kD; Tprg1l; Tprgl; transformation related protein 63 regulated like; tumor protein p63 regulated 1-like; tumor protein p63-regulated gene 1-like protein
Gene Symbols TPRG1L, Tprgl
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TPRG1L. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127262, 67808
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.