Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPRKB Antibody (OTI3H3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274574MFV450
Description
TPRKB Monoclonal antibody specifically detects TPRKB in Human, Mouse, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
TPRKB | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI3H3 | |
Flow Cytometry | |
CGI-121, PRPK (p53-related protein kinase)-binding protein, PRPK-binding protein, TP53RK binding protein, TP53RK-binding protein | |
Full length human recombinant protein of human TPRKB (NP_057142) produced in E.coli. | |
0.1 mL | |
Signal Transduction | |
51002 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction