Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPRKB Monoclonal Antibody (OTI1E9)
GREENER_CHOICE

Catalog No. PIMA526407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526407 Supplier Invitrogen™ Supplier No. MA526407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPRKB Monoclonal Antibody for Western Blot, IHC (P)

TPRKB is a component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t6A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. TPRKB acts as an allosteric effector that regulates the t6A activity of the complex. TPRKB is not required for tRNA modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPRKB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TPRKB
Gene Accession No. Q9Y3C4
Gene Alias 0610033G21Rik; 1810034M08Rik; CGI121; Cgi-121; DRWMS1; EKC/KEOPS complex subunit TPRKB; mDRWMS1; My019; Prpk (p53-related protein kinase)-binding protein; PRPK-binding protein; RGD1309786; Tp53rk binding protein; TP53RK-binding protein; TP53RK-binding protein; EKC/KEOPS complex subunit TPRKB; TPRKB; WBSCR18; zgc:101896
Gene Symbols TPRKB
Host Species Mouse
Immunogen Full length human recombinant protein of TPRKB produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51002
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.