Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPRKB Monoclonal Antibody (OTI3E3)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526380
Description
TPRKB Monoclonal Antibody for Western Blot, IHC (P), Flow
TPRKB is a component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t6A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. TPRKB acts as an allosteric effector that regulates the t6A activity of the complex. TPRKB is not required for tRNA modification.
Specifications
TPRKB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y3C4 | |
TPRKB | |
Full length human recombinant protein of TPRKB produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E3 | |
Unconjugated | |
TPRKB | |
0610033G21Rik; 1810034M08Rik; CGI121; Cgi-121; DRWMS1; EKC/KEOPS complex subunit TPRKB; mDRWMS1; My019; Prpk (p53-related protein kinase)-binding protein; PRPK-binding protein; RGD1309786; Tp53rk binding protein; TP53RK-binding protein; TP53RK-binding protein; EKC/KEOPS complex subunit TPRKB; TPRKB; WBSCR18; zgc:101896 | |
Mouse | |
Affinity chromatography | |
RUO | |
51002 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction