Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPRKB Mouse anti-Human, Clone: 2D5B9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TPRKB |
---|---|
Clone | 2D5B9 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6830
![]() ![]() |
Proteintech Group Inc
664021IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TPRKB is a component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t6A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. TPRKB acts as an allosteric effector that regulates the t6A activity of the complex. TPRKB is not required for tRNA modification.Specifications
TPRKB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CGI 121, My019, PRPK binding protein, TP53RK binding protein, TPRKB | |
TPRKB | |
IgG1 | |
Protein G | |
Antibody |
2D5B9 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9Y3C4 | |
51002 | |
TPRKB Fusion Protein Ag18770 | |
Primary | |
-20°C | |
TPRKB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title