Learn More
TPRKB Mouse anti-Human, Clone: OTI3E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800155

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TPRKB is a component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t6A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. TPRKB acts as an allosteric effector that regulates the t6A activity of the complex. TPRKB is not required for tRNA modification.Specifications
TPRKB | |
Monoclonal | |
Unconjugated | |
TPRKB | |
CGI-121, CGI121, GAMOS5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51002 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y3C4 | |
TPRKB | |
Full length human recombit protein of human TPRKB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.