Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPRKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544625 Supplier Thermo Scientific Supplier No. PA544625
Add to Cart
Add to Cart

TPRKB Polyclonal Antibody

TPRKB Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TPRKB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TPRKB
Gene Accession No. Q9Y3C4
Gene Alias 0610033G21Rik; 1810034M08Rik; CGI121; Cgi-121; DRWMS1; EKC/KEOPS complex subunit TPRKB; mDRWMS1; My019; Prpk (p53-related protein kinase)-binding protein; PRPK-binding protein; RGD1309786; Tp53rk binding protein; TP53RK-binding protein; TP53RK-binding protein; EKC/KEOPS complex subunit TPRKB; TPRKB; WBSCR18; zgc:101896
Gene Symbols TPRKB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TPRKB.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51002
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.