Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPRXL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569786
Description
This target displays homology in the following species: Human: 100%.
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This pseudogene is a member of the TPRX homeobox gene family.
Specifications
TPRXL | |
Polyclonal | |
Unconjugated | |
TPRXL | |
tetrapeptide repeat homeobox like; TPRX3P; TPRXL | |
Rabbit | |
Affinity chromatography | |
RUO | |
348825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q17RH7 | |
TPRXL | |
synthetic peptide directed towards the C terminal region of human TPRXL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction