Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tryptase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525355
Description
Tryptase Polyclonal Antibody for Western Blot, Flow
Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.
Specifications
| Tryptase | |
| Polyclonal | |
| Unconjugated | |
| Tpsab1 | |
| TPSG1/TPSB2/TPSAB1/TPSD1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q15661 | |
| Tpsab1 | |
| KLH conjugated synthetic peptide between 61-87 amino acids from the central region of human TPSAB1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction