Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TPSAB1 Recombinant Protein

Catalog No. 89968798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-798 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-798 Supplier Abnova™ Supplier No. H00007177P01
Add to Cart
Add to Cart

Used for ELISA, Western Blotting (Recombinant Protein), Antibody Production and Protein Array

Sequence: MLSLLLLALPVLASPAYAAPAPVQALQQAGIVGGQEAPRSKWPWQVSLRVRDRYWMHFCGGSLIHPQWVLTAAHCLGPDVKDLATLRVQLREQHLYYQDQLLPVSRIIVHPQFYIIQTGADIALLELEEPVNISSRVHTVMLPPASETFPPGMPCWVTGWGDVDNDEPLPPPFPLKQVKVPIMENHICDAKYHLGAYTGDDVRIIRDDMLCAGNTRRDSCQGDSGGPLVCKVNGTWLQAGVVSWDEGCAQPNRPGIYTRVTYYLDWIHHYVPKKP

Specifications

Accession Number AAH28059
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris HCl, 10 mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 7177
Molecular Weight (g/mol) 55.99 kDa
Name TPSAB1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Immunogen MLSLLLLALPVLASPAYAAPAPVQALQQAGIVGGQEAPRSKWPWQVSLRVRDRYWMHFCGGSLIHPQWVLTAAHCLGPDVKDLATLRVQLREQHLYYQDQLLPVSRIIVHPQFYIIQTGADIALLELEEPVNISSRVHTVMLPPASETFPPGMPCWVTGWGDVDNDEPLPPPFPLKQVKVPIMENHICDAKYHLGAYTGDDVRIIRDDMLCAGNTRRDSCQGDSGGPLVCKVNGTWLQAGVVSWDEGCAQPNRPGIYTRVTYYLDWIHHYVPKKP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias MCP7, TPS1, TPS2, TPSB1
Common Name TPSAB1
Gene Symbol TPSAB1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.