Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPSB2 Monoclonal Antibody (286820)
GREENER_CHOICE

Catalog No. PIMA523987
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523987 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523987 Supplier Thermo Scientific Supplier No. MA523987
Add to Cart
Add to Cart

TPSB2 Monoclonal Antibody (286820)

TPSB2 Monoclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen TPSB2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 286820
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Tpsb2
Gene Accession No. P21845
Gene Alias AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C
Gene Symbols Tpsb2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-6/Mcpt6 Ile32-Ser276.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17229
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.