Learn More
Invitrogen™ TPSB2 Monoclonal Antibody (349414)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TPS1, rhTPSG1, recombinant mouse (rm) TPS5, rmMCPT1, rmMCPT6, rmMCPT7, or rmMCPT11 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | TPSB2 |
| Applications | Immunoprecipitation, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 349414 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Tpsb2 |
| Gene Accession No. | P20231 |
| Gene Alias | AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.