Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPSB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119481
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119481 200 μL
PIPA5121533 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119481 Supplier Invitrogen™ Supplier No. PA5119481
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TPSB2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TPSB2 is a protein coding gene. Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. These genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. The alleles of this gene exhibit an unusual amount of sequence variation, such that the alleles were once thought to represent two separate genes, beta II and beta III. Beta tryptases appear to be the main isoenzymes expressed in mast cells, whereas in basophils, alpha-tryptases predominate. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPSB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Tpsb2
Gene Accession No. P50343
Gene Alias AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C
Gene Symbols Tpsb2
Host Species Rabbit
Immunogen Recombinant protein Tryptase Beta 2. The antigen corresponds to amino acid range 30-274 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29268
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.