Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPSD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24677625UL
Description
TPSD1 Polyclonal antibody specifically detects TPSD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TPSD1 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9BZJ3 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23430 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Delta-tryptase, EC 3.4.21, EC 3.4.21.59, HmMCP-3-like tryptase III, Mast cell mMCP-7-like, mast cell tryptase, MCP7L1, MCP7-LIKE, MGC95428, MMCP-7L, mMCP-7-like delta II tryptase, mMCP-7-like-1, mMCP-7-like-2, tryptase delta, tryptase delta 1, Tryptase-3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KDLAALRVQLREQHLYYQDQLLPVSRIIV | |
25 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction